Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01511

Sigma-Aldrich

Coumarin 153

BioReagent, suitable for fluorescence, ≥99.0% (HPLC)

Synonym(s):

2,3,6,7-Tetrahydro-9-(trifluoromethyl)-1H,5H,11H-[1]benzopyrano(6,7,8-ij)quinolizin-11-one, 2,3,6,7-Tetrahydro-9-trifluoromethyl-1H,5H-quinolizino(9,1-gh)coumarin, 8-Trifluoromethyl-2,3,5,6-4H-1,H-11-oxa-3a-aza-benzo[de]anthracen-10-one, Coumarin 540A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14F3NO2
CAS Number:
Molecular Weight:
309.28
Beilstein/REAXYS Number:
3624201
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

product line

BioReagent

assay

≥99.0% (HPLC)

mp

163-168 °C
164-168 °C (lit.)

λmax

422 nm

fluorescence

λem 532 nm in ethanol (Lasing peak 550 nm, lasing range 521 - 605 nm (methanol), pump source XeCl (308 nm))
λem 532 nm±5 nm

suitability

suitable for fluorescence

Storage temp.

2-8°C

SMILES string

FC(F)(F)C1=CC(=O)Oc2c3CCCN4CCCc(cc12)c34

InChI

1S/C16H14F3NO2/c17-16(18,19)12-8-13(21)22-15-10-4-2-6-20-5-1-3-9(14(10)20)7-11(12)15/h7-8H,1-6H2

Inchi Key

VSSSHNJONFTXHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Laser dye

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitaliy Kapishon et al.
Biomacromolecules, 16(7), 2040-2048 (2015-06-13)
Alginate-based amphiphilic graft copolymers were synthesized by single electron transfer living radical polymerization (SET-LRP), forming stable micelles during polymerization induced self-assembly (PISA). First, alginate macroinitiator was prepared by partial depolymerization of native alginate, solubility modification and attachment of initiator. Depolymerized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service