Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F1135

Sigma-Aldrich

Iron(III) sulfate hydrate

Fe 21-23 %

Synonym(s):

Ferric sulfate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe2(SO4)3 · xH2O
CAS Number:
Molecular Weight:
399.88 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

composition

Fe, 21-23%

SMILES string

[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Fe.3H2O4S/c;;3*1-5(2,3)4/h;;3*(H2,1,2,3,4)/q2*+3;;;/p-6

InChI key

RUTXIHLAWFEWGM-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Boateng et al.
Journal of applied microbiology, 111(6), 1533-1543 (2011-09-22)
To investigate the potential activation of hydrogen peroxide by a novel catalyst, reducing the concentration of hydrogen peroxide required and the time taken for microbial inactivation. The antimicrobial properties of an iron-based novel heterogeneous polyacrylonitrile catalyst in combination with hydrogen
P S Gillenwater et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 65(1), 112-118 (2011-12-17)
In this study, a broad range of readily deployable metal removal technologies were tested on a US refinery's wastewater to determine vanadium, arsenic and selenium removal performance. The bench-scale treatability studies were designed and performed so that test conditions could
Hanaa Shalan et al.
Quintessence international (Berlin, Germany : 1985), 43(6), 517-523 (2012-04-26)
To compare the effect of the pulpotomy medicaments glutaraldehyde, ferric sulfate, and formocresol on the structure and shear bond strength of a self-etch adhesive to the dentin of primary teeth. Forty human primary molars were sectioned mesiodistally and divided into
Homa Azargoon et al.
Journal of endodontics, 37(6), 807-811 (2011-07-27)
Obtaining hemostasis in the surgical crypt during periradicular surgery is essential. It allows for improved visibility and contributes to a dry environment suitable for the placement of moisture-sensitive root-end filling material. Although current materials may not be moisture sensitive during
M I Murav'ev et al.
Prikladnaia biokhimiia i mikrobiologiia, 47(6), 663-671 (2012-02-01)
Leaching of sulfide-oxidized copper concentrate of the Udokan deposit ore with a copper content of 37.4% was studied. In the course of treatment in a sulfuric acid solution with pH 1.2, a copper leaching rate was 6.9 g/kg h for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service