Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31272

Sigma-Aldrich

Potassium thiocyanate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%

Synonym(s):

Potassium rhodanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KSCN
CAS Number:
Molecular Weight:
97.18
Beilstein/REAXYS Number:
3594799
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99%
form:
crystals

grade

ACS reagent
puriss. p.a.

agency

USP/NF
reag. ISO
reag. Ph. Eur.

assay

≥99%

form

crystals

technique(s)

titration: suitable

impurities

≤0.001% ammonium (NH4)
≤0.005% water insoluble matter
≤0.01% insoluble in ethanol
≤0.025% matter consuming I (as I)

pH

5.3-8.5 (25 °C, 5%)

mp

173 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤10 mg/kg
sulfide (S2-): ≤10 mg/kg

cation traces

Cu: ≤2 mg/kg
Fe: ≤1 mg/kg
Na: ≤50 mg/kg
Pb: ≤2 mg/kg
heavy metals (as Pb): ≤5 mg/kg

SMILES string

[K]SC#N

InChI

1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1

InChI key

ZNNZYHKDIALBAK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Phase transition studies of potassium thiocyanate (KSCN) suggest that thiocyanate ions exhibit order-disorder type transitions.[1] Its polarized infrared spectrum has been reported.[2] It participates in the ring-opening of aziridines. This reaction was carried out in the presence of sulfated zirconia to afford the corresponding β-aminothiocyanates.[3]

Application

Potassium thiocyanate (KSCN) may be used to study the effect of emulsifier type on the micro-structural changes occurring in emulsified lipids during in vitro digestion.[4] It may be used for colorimetric quantification of intracellular iron content of superparamagnetic iron oxide (SPIO) nanoparticles.[5]

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

supp_hazards

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikolay A Semenov et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(49), 12983-12991 (2018-06-09)
Donor-acceptor (D-A) complexes between 3,4-dicyano-1,2,5-chalcogenadiazoles [chalcogen=Te (1 a), Se (1 b), S (1 c)] and the pseudohalides CN- and XCN- (X=O, S, Se, Te) were studied experimentally and theoretically. For 1 a, they were isolated as [K(18-crown-6)][1 a-CN] (2), [K(18-crown-6)][1 a-NCO] (3), [K(18-crown-6)][1 a-SCN] (4), [K(18-crown-6)][1 a-SeCN] (5)
Influence of initial emulsifier type on microstructural changes occurring in emulsified lipids during in vitro digestion.
Hur SJ, et al.
Food Chemistry, 114(1), 253-262 (2009)
Regioselective ring-opening of aziridines with potassium thiocyanate and thiols using sulfated zirconia as a heterogeneous recyclable catalyst.
Das B, et al.
Tetrahedron Letters, 47(5), 779-782 (2006)
The Phase Transition of Crystalline Potassium Thiocyanate, KSCN. II. X-Ray Study.
Yamada Y and Watanabe T.
Bulletin of the Chemical Society of Japan, 36(8), 1032-1037 (1963)
Polarized infrared spectrum of potassium thiocyanate.
Jones LH.
J. Chem. Phys. , 28(6), 1234-1236 (1958)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service