Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31236

Sigma-Aldrich

Iron(II) sulfate heptahydrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.0-103.4% (manganometric)

Synonym(s):

Ferrous sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeSO4 · 7H2O
CAS Number:
Molecular Weight:
278.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
99.0-103.4% (manganometric)
form:
crystals

grade

ACS reagent
puriss. p.a.

agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor pressure

14.6 mmHg ( 25 °C)

assay

99.0-103.4% (manganometric)

form

crystals

shelf life

Shelf life is limited

impurities

≤0.001% total nitrogen (N)

pH

3-4 (20 °C, 5%)

density

1.898 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤50 mg/kg
Cu: ≤10 mg/kg
Fe3+: ≤250 mg/kg
K: ≤20 mg/kg
Mg: ≤20 mg/kg
Mn: ≤500 mg/kg
Na: ≤20 mg/kg
Pb: ≤5 mg/kg
Zn: ≤20 mg/kg

SMILES string

O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

InChI

1S/Fe.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

Inchi Key

SURQXAFEQWPFPV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Packaging

bottled under inert gas

Caution

tends to oxidation by air

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal analysis of iron (II) sulphate heptahydrate in air. V: Thermal decomposition of hydroxy and oxysulphates.
Swamy M and Prasad T.
Journal of Thermal Analysis and Calorimetry, 20(1), 107-114 (1981)
Thermal analysis of iron (II) sulphate heptahydrate in air: III. Thermal decomposition of intermediate hydrates.
Swami MSR and Prasad TP.
Journal of Thermal Analysis and Calorimetry, 19(2), 297-304 (1980)
Hsiao-Chi Chuang et al.
Toxicology and applied pharmacology, 266(3), 329-334 (2012-12-25)
Pulmonary epithelial lining fluid (ELF) is the first substance to make contact with inhaled particulate matter (PM) and interacts chemically with PM components. The objective of this study was to determine the role of ELF in oxidative stress, DNA damage
Xiao-Kui Ma et al.
Carbohydrate polymers, 92(2), 1188-1196 (2013-02-13)
The influence of several components on exopolysaccharide (EPS) production and antioxidative activity (TEAC, Trolox-Equivalent Antioxidant Capacity) as well as their effects on the morphological development and cell viability of Phellinus sp. P0988 was determined. Rutin, FeSO(4), Tween 80 and complex
Roshini Ebenezer et al.
Tropical medicine & international health : TM & IH, 18(8), 942-951 (2013-07-11)
To assess the impact of deworming and iron supplementation on the cognitive abilities and educational achievement of school-age children in Sri Lanka. Prospective, placebo-controlled randomised study. The treatment group received deworming and weekly iron supplementation for 6 months; the control group

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service