Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13414

Sigma-Aldrich

Sodium carbonate decahydrate

puriss., meets analytical specification of Ph. Eur., BP, FCC, E500, 99.5-100.5% (ex dried subst.)

Synonym(s):

Sal soda, Soda

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3 · 10H2O
CAS Number:
Molecular Weight:
286.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

puriss.

assay

99.5-100.5% (ex dried subst.)

form

solid

quality

meets analytical specification of Ph. Eur., BP, FCC, E500

impurities

alkali hydroxides and-bicarbonates, complies

loss

55-65% loss on drying

anion traces

chloride (Cl-): ≤30 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Cu: ≤10 mg/kg
Fe: ≤5 mg/kg
Hg: ≤1 mg/kg
K: ≤200 mg/kg
Pb: ≤4 mg/kg
Zn: ≤10 mg/kg

suitability

complies for appearance of solution

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O

InChI

1S/CH2O3.2Na.10H2O/c2-1(3)4;;;;;;;;;;;;/h(H2,2,3,4);;;10*1H2/q;2*+1;;;;;;;;;;/p-2

InChI key

XYQRXRFVKUPBQN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical structure of poly (β-cyclodextrin-co-citric acid).
Bednarz S, et al.
Journal of Applied Polymer Science, 119(6), 3511-3520 (2011)
An infra-red study of the hydrates of sodium carbonate.
Buijs K and Schutte CJH.
Spectrochimica Acta Part A: Molecular Spectroscopy, 17(9), 917-920 (1961)
Dehydration of sodium carbonate decahydrate to monohydrate in a fluidized bed.
Hartman M, et al.
AIChE Journal, 47(10), 2333-2333 (2001)
Azadeh Nilghaz et al.
Lab on a chip, 12(1), 209-218 (2011-11-18)
This paper describes the fabrication of microfluidic cloth-based analytical devices (μCADs) using a simple wax patterning method on cotton cloth for performing colorimetric bioassays. Commercial cotton cloth fabric is proposed as a new inexpensive, lightweight, and flexible platform for fabricating
Dries Vandamme et al.
Bioresource technology, 105, 114-119 (2011-12-21)
Microalgae hold great potential as a feedstock for biofuels or bulk protein or treatment of wastewater or flue gas. Realising these applications will require the development of a cost-efficient harvesting technology. Here, we explore the potential of flocculation induced by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service