Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06208

Supelco

Sodium hydroxide

pellets, Cl <0.001%

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

assay

98-102% HCl basis (titration)

form

pellets

impurities

≤10 mg/kg heavy metals (as Ag)

mp

318 °C (lit.)

solubility

water: soluble 1260 g/L at 20 °C

density

2.13 g/cm3

anion traces

chloride (Cl-): ≤10 mg/kg

cation traces

heavy metals (as Ag): ≤10 mg/kg

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemoselective, solvent-free aldol condensation reaction.
Raston CL and Scott JL.
Green Chemistry, 2(2), 49-52 (2000)
M A Roberts et al.
Analytical chemistry, 69(11), 2035-2042 (1997-06-01)
This report describes a UV laser photoablation method for the production of miniaturized liquid-handling systems on polymer substrate chips. The fabrication of fluid channel and reservoir networks is accomplished by firing 200 mJ pulses from an UV excimer laser at
Silvio Carrettin et al.
Chemical communications (Cambridge, England), (7)(7), 696-697 (2002-07-18)
Glycerol is oxidised to glyceric acid with 100% selectivity using either 1% Au/charcoal or 1% Au/graphite catalyst under mild reaction conditions (60 degrees C, 3 h, water as solvent).
Determination of specific surface area of colloidal silica by titration with sodium hydroxide.
Sears Jr GW.
Analytical Chemistry, 28(12), 1981-1983 (1956)
M Chevallet et al.
Electrophoresis, 19(11), 1901-1909 (1998-09-18)
Severe quantitative loss of protein is often observed in high-resolution two-dimensional electrophoresis of membrane proteins, while the resolution is usually not affected. To improve the solubility of proteins in this technique, we tested denaturing cocktails containing various detergents and chaotropes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service