Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SK013

Supelco

VOC Calibration Verification

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

Quality Level

format

multi-component solution

Storage temp.

15-25°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    2-Butoxyethanol 9.00 % w/v
    n-Octane 9.00 % w/v
    Acetone 9.00 % w/v
    n-Butyl alcohol 9.00 % w/v
    1,2-Dichlorobenzene 9.00 % w/v
    Dichloromethane 9.00 % w/v
    Ethyl acetate 9.00 % w/v
    Methanol 6.66 % w/v
    Toluene 10.0 % w/v
    1,1,1-Trichloroethane 9.00 % w/v
    n-Hexadecane 3.00 % w/v
    See All (11)

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Ozone 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 2 - STOT SE 3

target_organs

Central nervous system, Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

69.8 °F

flash_point_c

21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service