Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5156

Supelco

Pyrithioxin dihydrochloride

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2O4S2 · 2HCl
CAS Number:
Molecular Weight:
441.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

vitamins, nutraceuticals, and natural products

format

neat

storage temp.

2-8°C

SMILES string

OC1=C(CO)C(CSSCC2=C(CO)C(O)=C(C)N=C2)=CN=C1C.Cl.Cl

InChI

1S/C16H20N2O4S2.2ClH/c1-9-15(21)13(5-19)11(3-17-9)7-23-24-8-12-4-18-10(2)16(22)14(12)6-20;;/h3-4,19-22H,5-8H2,1-2H3;2*1H

InChI key

HRZWYHGGJOWGRI-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Occupational contact dermatitis due to pyritinol.
W Wigger-Alberti et al.
Contact dermatitis, 37(2), 91-92 (1997-08-01)
M K C Nair et al.
Indian pediatrics, 46 Suppl, s37-s42 (2009-05-16)
To evaluate the efficacy of pyritinol in improving the neurodevelopmental outcome at one year of age among term babies with post-asphyxial encephalopathy. Level II Neonatal Nursery and Child Development Centre, Medical College, Thiruvananthapuram. Randomised placebo controlled double blind trial. 108
Guillermina Yanek Jiménez-Andrade et al.
European journal of pharmacology, 590(1-3), 170-176 (2008-07-03)
The purpose of this study was to assess the antinociceptive and antiallodynic effect of pyritinol as well as its possible mechanism of action in diabetic rats. Streptozotocin (50 mg/kg) injection caused hyperglycemia within 1 week. Formalin-evoked flinching was increased in
H Hartmann et al.
Neuropharmacology, 32(2), 119-125 (1993-02-01)
The effect of pyritinol, a commonly used nootropic drug, on receptor properties and function was investigated in different neuronal systems, possibly associated with age-related decline in brain function. Chronic treatment (15 days) of aged (22 months) female NMRI mice with
N N Zavadenko et al.
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova, 112(7 Pt 2), 90-95 (2013-01-19)
To assess the efficacy of treatment with encephabol, we examined 40 children, aged from 3 to 5 years, with developmental dysphasia. All patients were randomized into two equal groups: group 1 received encephabol (suspension form, daily dosage 200-250 mg, or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service