Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST8535A

VSMOW2 Vienna Standard Mean Ocean Water 2 (Hydrogen and Oxygen Isotopes in Water)

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

isotopic purity

 2H
 18O

form

liquid

packaging

pkg of 20 mL

manufacturer/tradename

NIST®

storage temp.

15-25°C

General description

This Reference Material (RM) is an international measurement standard that defines the zero of the VSMOW-SLAP (Vienna Standard Mean Ocean Water-Standard Light Antarctic Precipitation) d-scales for isotope-number ratios of hydrogen (H) and oxygen (O). The equivalent name for this RM, as used by the International Atomic Energy Agency (IAEA) and the U.S. Geological Survey (USGS), is VSMOW2. A unit of RM 8535a consists of one ampoule containing approximately 20 mL of water.
SRM 8535a_cert SRM 8535a _SDS

Other Notes

Reference/Informational value is provided for the following analytes.
Hydrogen (1H/2H), Oxygen (16O/18O)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service