Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1548A

Typical diet

NIST® SRM® 1548a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of set (2)

manufacturer/tradename

NIST®

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

General description

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Ash, Calories, Carbohydrates, Total Dietary Fiber, Total Fat, Nitrogen, Phosphorous, Protein
Trace Elements: Aluminum (Al), Antimony (SB), Arsenic (As), Barium (Ba), Boron (B), Bromine (Br), Cadmium (Cd), Calcium (Ca), Cesium (Cs), Chlorine (Cl2), Cobalt (Co), Copper (Cu), Iodine (I), Iron (Fe), Lead (Pb), Magnesium (Mg), Manganese (Mn), Mercury (Hg), Molybdenum (Mo), Nickel (Ni), Potassium (K), Scandium (Sc), Selenium (Se), Silicon (Si), Sodium (Na), Strontium (Sr), Sulfur (S), Tin (Sn), Titanium (Ti), Zinc (Zn)

Matrix Group: Processed Food

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service