Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM646

Sulfur-34S solution

IRMM®, certified reference material, Spike Isotopic Reference Material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
34S
CAS Number:
Molecular Weight:
33.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

agency

IRMM®

manufacturer/tradename

JRC

concentration

4586 μmol/kg±27 μmol/kg 34S in 2.8 M HNO3

application(s)

pharmaceutical (small molecule)

format

single component solution

SMILES string

[34S]

InChI

1S/S/i1+2

InChI key

NINIDFKCEFEMDL-NJFSPNSNSA-N

Analysis Note

For more information please see:
IRMM646

Legal Information

IRMM is a registered trademark of European Commission

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service