Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM469

3,3′,5 Triiodothyronine (T3)

IRMM®, certified reference material

Synonym(s):

3,3′,5-Triiodo-L-thyronine, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine, Liothyronine, T3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12I3NO4
CAS Number:
Molecular Weight:
650.97
Beilstein/REAXYS Number:
2710227
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material

agency

IRMM®

mp

234-238 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−70°C

SMILES string

N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C(O)=O

InChI

1S/C15H12I3NO4/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22/h1-4,6,12,20H,5,19H2,(H,21,22)/t12-/m0/s1

InChI key

AUYYCJSJGJYCDS-LBPRGKRZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
IRMM469

Legal Information

IRMM is a registered trademark of European Commission

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1

target_organs

Thyroid,Cardio-vascular system,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ben Emery et al.
Cell, 138(1), 172-185 (2009-07-15)
The transcriptional control of CNS myelin gene expression is poorly understood. Here we identify gene model 98, which we have named myelin gene regulatory factor (MRF), as a transcriptional regulator required for CNS myelination. Within the CNS, MRF is specifically
J Himms-Hagen
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 4(11), 2890-2898 (1990-08-01)
Energy expenditure for thermogenesis in brown adipose tissue (BAT) serves either to maintain body temperature in the cold or to waste food energy. It has roles in thermal balance and energy balance, and when defective, is usually associated with obesity.
E Gunilla Stalenheim
Psychiatry research, 121(3), 281-291 (2003-12-17)
This study is a follow-up investigation of a forensic psychiatric sub-population 6-8 years after forensic psychiatric evaluation. The aim was to examine the long-term validity of biological markers of psychopathy and antisocial behavior over time. Data on criminal records were
R Calvo et al.
The Journal of clinical investigation, 86(3), 889-899 (1990-09-01)
To study the protective effects of maternal thyroxine (T4) and 3,5,3'-triiodothyronine (T3) in congenital hypothyroidism, we gave pregnant rats methimazole (MMI), an antithyroid drug that crosses the placenta, and infused them with three different doses of T4 or T3. The
Nathan A Lack et al.
Journal of medicinal chemistry, 54(24), 8563-8573 (2011-11-04)
The androgen receptor (AR) is the best studied drug target for the treatment of prostate cancer. While there are a number of drugs that target the AR, they all work through the same mechanism of action and are prone to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service