Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E0530000

4-Epioxytetracycline

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O9
CAS Number:
Molecular Weight:
460.43
UNSPSC Code:
41116107

application(s)

pharmaceutical (small molecule)

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 40 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

pictograms

Health hazardEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aytekin Çelik et al.
Journal of hazardous materials, 356, 26-33 (2018-05-29)
Pharmaceuticals in waterbodies are a growing concern due to their extensive uses and adverse effects on aquatic life. Oxytetracycline (OTC) is one of tetracycline antibiotic group used for treatment of animals and humans. This study evaluates the simultaneous oxidation of
A L Stanton et al.
Journal of dairy science, 95(9), 4950-4960 (2012-08-25)
Bovine respiratory disease complex (BRD) is a common disease in weaned dairy calves that incurs economic and welfare costs. This study was an extension of a randomized clinical trial in which a single injection of tulathromycin (TUL) or oxytetracycline (TET)
Leslie R Wintz et al.
American journal of veterinary research, 73(12), 1951-1956 (2012-11-28)
To describe the effect of systemically administered oxytetracycline on the viscoelastic properties of rat tail tendon fascicles (TTfs) to provide a mechanistic rationale for pharmacological treatment of flexural limb deformities in foals. TTfs from ten 1-month-old and ten 6-month-old male
Guang-Feng Yang et al.
Bioresource technology, 129, 65-71 (2012-12-13)
The short- and long-term effects of oxytetracycline (OTC) on the anaerobic ammonium oxidation (Anammox) process were evaluated. The OTC inhibition of Anammox was substrate-, and especially nitrite-, dependent. The IC50 of OTC in the batch tests on an Anammox mixed
Gregory J Barord et al.
Journal of zoo and wildlife medicine : official publication of the American Association of Zoo Veterinarians, 43(3), 636-639 (2012-10-23)
A single Nautilus pompilius manifested a bacterial infection and nematode infestation soon after it was received from the wild, resulting in a significant buildup of mucus above the left eye and tentacles. This condition is known to lead to rapid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service