Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D5761

Supelco

5,5-Diallylbarbituric acid

analytical standard

Synonym(s):

Allobarbital

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O3
CAS Number:
Molecular Weight:
208.21
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

drug control

USDEA Schedule III; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

C=CCC1(CC=C)C(=O)NC(=O)NC1=O

InChI

1S/C10H12N2O3/c1-3-5-10(6-4-2)7(13)11-9(15)12-8(10)14/h3-4H,1-2,5-6H2,(H2,11,12,13,14,15)

Inchi Key

FDQGNLOWMMVRQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Sedative/hypnotic

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Kojima et al.
International journal of legal medicine, 107(1), 21-24 (1994-01-01)
A simple and rapid method for analysis of barbiturates in serum has been developed. In order to extract and clean barbiturates in serum, a separation column packed with Extrelut and Florisil was used, and the eluate was directly analyzed by
Z Song et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 130(2), 237-250 (2001-09-07)
This study was conducted to investigate the hepatic extraction of plasma free amino acids in anesthetized Single Comb White Leghorn (SCWL) males (Gallus domesticus). SCWL males were anesthetized and implanted with cannulae in the carotid artery, hepatic vein, hepatic portal
S Furukawa et al.
The Journal of veterinary medical science, 62(1), 23-28 (2000-02-17)
To examine the effect on cell population in hepatocytes of phenobarbital (PB) and other barbiturates, PB, allobarbital (ALB), barbital sodium (BS) and barbituric acid (BA) were given orally to male rats for 7 consecutive days. Although there was no apparent
Y Martín-Biosca et al.
Journal of pharmaceutical and biomedical analysis, 21(2), 331-338 (2000-03-07)
A liquid chromatographic procedure for the determination of six barbiturates (barbital, diallyl barbituric acid, phenobarbital, butabarbital, amobarbital and pentobarbital) in urine samples is described. The proposed system uses a Spherisorb octadecyl-silane ODS-2 C18 analytical column and a guard column of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service