Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM8009

Supelco

Ultra Pure Nitric Acid, 2%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

packaging

bottle of 125 mL

Quality Level

storage temp.

room temp

General description

Our ultra pure 2% nitric acid product may be used for dilution or to establish base lines. The calibration, reagent, and rinse blanks are prepared by diluting the appropriate acid with water and any necessary internal standards to produce the required acid concentration, generally 1% HNO3.

pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service