Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM055

Supelco

Trace Metals - Sewage Sludge 4

certified reference material, pkg of 50 g (dry)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

CofA

current certificate can be downloaded

composition

Chemical oxygen demand (COD)

packaging

pkg of 50 g (dry)

manufacturer/tradename

RTC CRM055-050

technique(s)

AAS: suitable
ICP: suitable

particle size

0.2-1 mm

application(s)

environmental

format

matrix material

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

The following elements are not certified and are included for information only:
  • Be, K, Se, Na
Values of analytes vary lot to lot. Some lots may not contain all analytes.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Analyte

Description

    Aluminum
    Antimony
    Arsenic
    Barium
    Beryllium
    Cadmium
    Calcium
    Chromium
    Cobalt
    Copper
    Iron
    Lead
    Magnesium
    Manganese
    Mercury
    Molybdenum
    Nickel
    Potassium
    Selenium
    Silver
    Sodium
    Vanadium
    Zinc
    See All (23)

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1A - Carc. 1B Inhalation - Muta. 1B - Skin Sens. 1 - STOT RE 2

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnieszka Medyńska-Juraszek et al.
International journal of environmental research and public health, 17(21) (2020-10-31)
Municipal green-waste compost and wheat straw biochar amendments were assessed for their assistance in regulating the mobility of Cu, Pb, Zn, Cd, Cr and Ni and the uptake of these metals by five commonly grown green leafy vegetables (radish, lettuce
Agnieszka Medyńska-Juraszek et al.
Materials (Basel, Switzerland), 13(11) (2020-06-03)
There is an urgent need to search for new sorbents of pollutants presently delivered to the environment. Recently biochar has received much attention as a low-cost, highly effective heavy metal adsorbent. Biochar has been identified as an efficient material for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service