Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B4144

Supelco

Bromazepam

analytical standard

Synonym(s):

7-Bromo-5-(2-pyridyl)-3H-1,4-benzodiaxepin-2(1H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10BrN3O
CAS Number:
Molecular Weight:
316.15
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

drug control

USDEA Schedule IV; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

O=C(CN=C1C2=CC=CC=N2)NC3=C1C=C(Br)C=C3

InChI

1S/C14H10BrN3O/c15-9-4-5-11-10(7-9)14(17-8-13(19)18-11)12-3-1-2-6-16-12/h1-7H,8H2,(H,18,19)

InChI key

VMIYHDSEFNYJSL-UHFFFAOYSA-N

Gene Information

human ... GABRA2(2555)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Anxiolytic

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Djezzar et al.
International journal of legal medicine, 123(3), 213-219 (2008-10-18)
Psychoactive substances may be administered without the knowledge of a victim in order to induce incapacitation and thus facilitate criminal actions. The characteristics of the victims and the drugs used in such suspected chemical submissions (CS) were analyzed in 309
M S Attia
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 74(4), 972-976 (2009-09-29)
A rapid, simple and sensitive spectrofluorimetric method for determination of trace amount of bromazepam is developed. In phosphate buffer of pH 7.4. The bromazepam enhance the luminescence intensity of the Eu(3+) ion in Eu(3+)-bromazepam complex at lambda(ex)=390nm. The produced luminescence
Marlo Cunha et al.
Neuroscience letters, 448(1), 33-36 (2008-10-22)
The aim of this study was to investigate the influence of bromazepam on EEG and the motor learning process when healthy subjects were submitted to a typewriting task. We investigated bromazepam due to its abuse by various populations and its
Yvan Gaillard et al.
Forensic science international, 207(1-3), e16-e18 (2011-01-05)
We reported on the death by poisoning of a one-month-old baby that had followed the death of one of her sister (due to cyamemazine overdose). Exhumation of the corpse was done 8 months after burial and revealed the presence of
Julio G Silva et al.
CNS & neurological disorders drug targets, 10(8), 945-950 (2012-01-11)
This study aimed to investigate the acute modulatory effect of bromazepam, a benzodiazepine derivative drug, on alpha and beta bands (8-35Hz) in primary motor areas (M1) through event-related spectral perturbation (ERSP). Ten healthy subjects were submitted to a cross-over double-blind

Questions

  1. Do you guys carry bromazepam 6 mg tablets?

    1 answer
    1. This product is not offered in tableted form. Bromazepam is considered a controlled substance and all products available are intended for research and analytical use only. They are not suitable for human use.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service