Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6138

Sigma-Aldrich

Agarose

Type III, High EEO

Synonym(s):

3,6-Anhydro-α-L-galacto-β-D-galactan, Agarose HE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

algae (marine)

type

Type III

form

powder

impurities

≤10% water

EEO

0.23-0.26

transition temp

gel point 36 °C ±1.5 °C

gel strength

≥650 g/cm2 (1% gel)

anion traces

sulfate (SO42-): ≤0.20%

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

The following is a list of properties associated with our agaroses:
Sulfate content - used as an indicator of purity, since sulfate is the major ionic group present.
Gel strength - the force that must be applied to a gel to cause it to fracture.
Gel point - the temperature at which an aqueous agarose solution forms a gel as it cools. Agarose solutions exhibit hysteresis in the liquid-to-gel transition - that is, their gel point is not the same as their melting temperature.
Electroendosmosis (EEO) - a movement of liquid through the gel. Anionic groups in an agarose gel are affixed to the matrix and cannot move, but dissociable counter cations can migrate toward the cathode in the matrix, giving rise to EEO. Since electrophoretic movement of biopolymers is usually toward the anode, EEO can disrupt separations because of internal convection.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethan G McBride et al.
Current biology : CB, 29(10), 1592-1605 (2019-05-06)
Sensory selection and movement locally and globally modulate neural responses in seemingly similar ways. For example, locomotion enhances visual responses in mouse primary visual cortex (V1), resembling the effects of spatial attention on primate visual cortical activity. However, interactions between
Maria Carmen Piñon et al.
The Journal of physiology, 587(Pt 9), 1903-1915 (2009-03-18)
In the Golli-tau-eGFP (GTE) transgenic mouse the reporter gene expression is largely confined to the layer of subplate neurons (SPn), providing an opportunity to study their intracortical and extracortical projections. In this study, we examined the thalamic afferents and layer
Víctor Carriel et al.
Journal of neural engineering, 10(2), 026022-026022 (2013-03-27)
The objective was to study the effectiveness of a commercially available collagen conduit filled with fibrin-agarose hydrogels alone or with fibrin-agarose hydrogels containing autologous adipose-derived mesenchymal stem cells (ADMSCs) in a rat sciatic nerve injury model. A 10 mm gap
P Upcroft et al.
Journal of chromatography, 618(1-2), 79-93 (1993-08-25)
Agarose as a medium for separation of DNA was first introduced in 1962 and since the early 1970s agarose submarine gel electrophoresis has been synonymous with separations of DNA molecules larger than 1 kilobase pair (kb). The large pore size
Julia Dahlmann et al.
Biomaterials, 34(10), 2463-2471 (2013-01-22)
In most pluripotent stem cell differentiation protocols the formation of embryoid bodies (EBs) is an important step. Here we describe a rapid, straightforward soft lithography approach for the preparation of hydrophilic silicon masters from different templates and the subsequent production

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service