Key Documents
99112
Sigma-Aldrich
Outlet valve No 70688
stainless steel, pkg of 1 ea
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
material
stainless steel
packaging
pkg of 1 ea
General description
Control valves for cylinders in stainless steel, for corrosive gases, angled, with nut at inlet and unscrewable hose outlet.
Inlet: W 21.8 mm x 14; left-hand thread Hose outlet: G1/4 in. (M).
Choose from one of the most recent versions:
Certificates of Analysis (COA)