Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94380

Supelco

tert-Butyl(chloro)diphenylsilane

for GC derivatization, LiChropur, ≥98.0% (GC)

Synonym(s):

tert-Butyldiphenylchlorosilane, TBDPSCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(C6H5)2Cl
CAS Number:
Molecular Weight:
274.86
Beilstein/REAXYS Number:
644023
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

assay

≥98.0% (GC)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

refractive index

n20/D 1.566-1.568
n20/D 1.568 (lit.)

bp

90 °C/0.01 mmHg (lit.)

density

1.057 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)[Si](Cl)(c1ccccc1)c2ccccc2

InChI

1S/C16H19ClSi/c1-16(2,3)18(17,14-10-6-4-7-11-14)15-12-8-5-9-13-15/h4-13H,1-3H3

InChI key

MHYGQXWCZAYSLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service