Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90061

Supelco

4-Deoxy-D-erythronic acid sodium salt hydrate

≥97.0% (GC)

Synonym(s):

(2R,3R)-2,3-Dihydroxybutyric acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O4 · xNa+ · yH2O
Molecular Weight:
120.10 (anhydrous free acid basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

Quality Level

assay

≥97.0% (GC)

optical activity

[α]/D -10±2°, c = 0.5 in 0.1 M NaOH

application(s)

clinical testing

format

neat

storage temp.

2-8°C

Biochem/physiol Actions

4-Deoxyerythronic acid has been associated with uremia and diabetes mellitus type 1, but is also a normal organic acid present in human biofluids, derived presumably from L-threonine.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F W Bultitude et al.
Clinical chemistry, 21(9), 1329-1334 (1975-08-01)
We describe a method for comparing plasma samples from healthy subjects and from chronic uremic patients before and after dialysis. It was used to determine the nature of those metabolites that appear to characterize the uremic state. Preliminary fractionation of
Konstantinos A Kouremenos et al.
Journal of chromatography. A, 1217(1), 104-111 (2009-11-10)
Comprehensive two-dimensional gas chromatography (GCxGC) time-of-flight mass spectrometry (ToFMS) was applied to the analysis of urinary organic acids from patients with inborn errors of metabolism. Abnormal profiles were obtained from all five patients studied. Methylmalonic academia and deficiencies of 3-methylcrotonyl-CoA
Sílvia O Diaz et al.
Journal of proteome research, 12(2), 969-979 (2012-12-13)
In this work, untargeted NMR metabonomics was employed to evaluate the effects of pregnancy on the metabolite composition of maternal urine, thus establishing a control excretory trajectory for healthy pregnancies. Urine was collected for independent groups of healthy nonpregnant and
D B Kassel et al.
Biomedical & environmental mass spectrometry, 13(10), 535-540 (1986-10-01)
Metabolic profiling of urinary organic acids from patients with juvenile-onset (Type 1) diabetes mellitus have revealed significantly elevated levels of 2-hydroxybutyric and 4-deoxythreonic acids. To test the hypothesis that these metabolites, as well as 4-deoxyerythronic acid, are derived from L-threonine
Yi Zhang et al.
Molecular cell, 73(6), 1127-1137 (2019-02-18)
We have previously proposed that selective inheritance, the limited transmission of damaging mtDNA mutations from mother to offspring, is based on replication competition in Drosophila melanogaster. This model, which stems from our observation that wild-type mitochondria propagate much more vigorously

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service