Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

89948

Supelco

Tectoridin

analytical standard

Synonym(s):

D-Glucopyranosyloxy)-5-hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, Shekanin, Tectorigenin 7-O-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22O11
CAS Number:
Molecular Weight:
462.40
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](OC2=CC(OC(C3=CC=C(O)C=C3)=CC4=O)=C4C(O)=C2OC)O[C@H](CO)[C@H]1O

InChI

1S/C22H22O11/c1-30-21-14(32-22-20(29)19(28)17(26)15(8-23)33-22)7-13-16(18(21)27)11(25)6-12(31-13)9-2-4-10(24)5-3-9/h2-7,15,17,19-20,22-24,26-29H,8H2,1H3/t15-,17-,19+,20-,22-/m1/s1

InChI key

GCLAFEGUXXHIFT-IWLDQSELSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyoung Ah Kang et al.
European journal of pharmacology, 519(1-2), 16-23 (2005-08-17)
In the present study, the antioxidative properties of tectorigenin, a metabolite formed by transformation of tectoridin by intestinal microflora, were investigated. Tectorigenin was found to scavenge intracellular reactive oxygen species, and 1,1-diphenyl-2-picrylhydrazyl (DPPH) radical, and thus prevented lipid peroxidation. The
Yong Chen et al.
The Journal of pharmacy and pharmacology, 60(6), 709-716 (2008-05-24)
In this work, liquid chromatography-electrospray ionization tandem ion-trap mass spectrometry (LC-MS(n)) was used to investigate the in-vivo and in-vitro metabolism of tectoridin. After oral administration of a single dose (100 mg kg(-1)) of tectoridin to healthy rats, faeces and urine
JangJa Hong et al.
Inflammation & allergy drug targets, 7(3), 195-202 (2008-09-11)
Effects of compounds isolated from medicinal plants in Korea on prostaglandin E2 (PGE2) production in rat peritoneal macrophages were examined, and mechanism of action of the active constituents was analyzed. The active constituents were as follows; tectorigenin and tectoridin isolated
Cheng-Feng Luo et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(19), 1497-1501 (2011-04-23)
A highly sensitive and specific method of rapid resolution liquid chromatography tandem mass spectrometry (RRLC-MS/MS) in positive ionization mode has been developed and validated for pharmacokinetic study of puerarin in rat plasma. Chromatography was carried out on a Zorbax XDB
Kyungsu Kang et al.
Molecules and cells, 27(3), 351-357 (2009-03-28)
Phytoestrogens are the natural compounds isolated from plants, which are structurally similar to animal estrogen, 17beta-estradiol. Tectoridin, a major isoflavone isolated from the rhizome of Belamcanda chinensis. Tectoridin is known as a phytoestrogen, however, the molecular mechanisms underlying its estrogenic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service