Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88950

Sigma-Aldrich

Thionyl chloride

puriss., ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SOCl2
CAS Number:
Molecular Weight:
118.97
Beilstein/REAXYS Number:
1209273
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor pressure

97 mmHg ( 20 °C)

grade

puriss.

assay

≥99.0%

form

liquid

color

APHA: ≤50

refractive index

n20/D 1.518 (lit.)

bp

79 °C (lit.)

mp

−105 °C (lit.)

density

1.631 g/mL at 25 °C (lit.)

SMILES string

ClS(Cl)=O

InChI

1S/Cl2OS/c1-4(2)3

InChI key

FYSNRJHAOHDILO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thionyl chloride is a sulfur containing acid halide.[1] It participates in the preparation of butadiyne (diacetylene).[2] On reaction with benzaldehyde, it affords benzylidene dichloride.[3] It is widely employed for the transformation of alcohols into reactive alkyl chlorides.[4]

Application

Thionyl chloride may be employed for the sulfonation of aromatic compounds.[5] It may be used in the synthesis of following:
  • cyclooctasulfur oxide[6]
  • disulfuroxide[6]
  • acetylchloride[1]
  • organochlorine compounds[7]
  • G-series nerve agents such as phosgene or chloropicrine[7]
  • 1-(chloromethyl)-3,5-dimethylbenzene[8]

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng Zhou et al.
Molecules (Basel, Switzerland), 20(6), 10122-10130 (2015-06-04)
Pincer (Phebox)Ir(mesityl)(OAc) (2) (Phebox = 3,5-dimethylphenyl-2,6-bis(oxazolinyl)) complex, formed by benzylic C-H activation of mesitylene (1,3,5-trimethylbenzene) using (Phebox)Ir(OAc)2OH2 (1), was treated with thionyl chloride to rapidly form 1-(chloromethyl)-3,5-dimethylbenzene in 50% yield at 23 °C. A green species was obtained at the
Eagleson M.
Concise Encyclopedia Chemistry, 1056-1056 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 118-118 (1994)
Jinjian Zheng et al.
Analytica chimica acta, 891, 255-260 (2015-09-22)
Thionyl chloride is often used to convert alcohols into more reactive alkyl chloride, which can be easily converted to many compounds that are not possible from alcohols directly. One important reaction of alkyl chloride is nucleophilic substitution, which is typically
Eagleson M.
Concise Encyclopedia Chemistry, 1049-1049 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service