Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73763

Supelco

Nicotinic acid

for inorganic trace analysis, ≥99.999% (metals basis)

Synonym(s):

3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid, Vitamin B3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein/REAXYS Number:
109591
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

Quality Level

assay

≥99.5% (HPLC)
≥99.999% (metals basis)

form

powder

color

colorless to white

mp

236-239 °C (lit.)

cation traces

Ag: ≤0.02 mg/kg
Al: ≤0.1 mg/kg
As: ≤0.05 mg/kg
Au: ≤0.01 mg/kg
Ba: ≤0.05 mg/kg
Be: ≤0.01 mg/kg
Bi: ≤0.01 mg/kg
Ca: ≤2 mg/kg
Cd: ≤0.01 mg/kg
Ce: ≤0.01 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.2 mg/kg
Cs: ≤0.01 mg/kg
Cu: ≤0.02 mg/kg
Dy: ≤0.01 mg/kg
Er: ≤0.01 mg/kg
Eu: ≤0.01 mg/kg
Fe: ≤0.5 mg/kg
Ga: ≤0.01 mg/kg
Gd: ≤0.01 mg/kg
Ge: ≤0.01 mg/kg
Hf: ≤0.01 mg/kg
Hg: ≤0.005 mg/kg
Ho: ≤0.01 mg/kg
In: ≤0.01 mg/kg
Ir: ≤0.01 mg/kg
K: ≤0.2 mg/kg
La: ≤0.01 mg/kg
Li: ≤0.01 mg/kg
Lu: ≤0.01 mg/kg
Mg: ≤0.05 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.01 mg/kg
Na: ≤0.5 mg/kg
Nb: ≤0.01 mg/kg
Nd: ≤0.01 mg/kg
Ni: ≤0.05 mg/kg
Os: ≤0.01 mg/kg
Pb: ≤0.01 mg/kg
Pd: ≤0.01 mg/kg
Pr: ≤0.01 mg/kg
Pt: ≤0.05 mg/kg
Rb: ≤0.01 mg/kg
Re: ≤0.01 mg/kg
Rh: ≤0.01 mg/kg
Ru: ≤0.01 mg/kg
Sb: ≤0.01 mg/kg
Sc: ≤0.05 mg/kg
Se: ≤0.05 mg/kg
Sm: ≤0.01 mg/kg
Sn: ≤0.01 mg/kg
Sr: ≤0.01 mg/kg
Ta: ≤0.01 mg/kg
Tb: ≤0.01 mg/kg
Te: ≤0.01 mg/kg
Th: ≤0.01 mg/kg
Ti: ≤0.05 mg/kg
Tl: ≤0.01 mg/kg
Tm: ≤0.01 mg/kg
U: ≤0.01 mg/kg
V: ≤0.01 mg/kg
W: ≤0.01 mg/kg
Y: ≤0.01 mg/kg
Yb: ≤0.01 mg/kg
Zn: ≤0.1 mg/kg
Zr: ≤0.01 mg/kg

SMILES string

OC(=O)c1cccnc1

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)

InChI key

PVNIIMVLHYAWGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinic acid is a water soluble B complex vitamin,[1] with effective plasma lipid-lowering effect and hence is clinically used to treat hyperlipidemia.[2]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

379.4 °F - closed cup

flash_point_c

193 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Molecular identification of nicotinic acid receptor
Soga T, et al.
Biochemical and biophysical research communications, 303(1), 364-369 (2003)
Sorin Tunaru et al.
Nature medicine, 9(3), 352-355 (2003-02-04)
Nicotinic acid (niacin), a vitamin of the B complex, has been used for almost 50 years as a lipid-lowering drug. The pharmacological effect of nicotinic acid requires doses that are much higher than those provided by a normal diet. Its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service