Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67653

Supelco

L-Kynurenine

analytical standard

Synonym(s):

β-Anthraniloyl-L-alanine, L-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O3
CAS Number:
Molecular Weight:
208.21
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

assay

≥98% (HPLC)

optical activity

[α]25/D -32.5 to -28.0°, c = 0.4 in H2O

shelf life

limited shelf life, expiry date on the label

impurities

≤5.0% water

application(s)

clinical testing

format

neat

SMILES string

N[C@@H](CC(=O)c1ccccc1N)C(O)=O

InChI

1S/C10H12N2O3/c11-7-4-2-1-3-6(7)9(13)5-8(12)10(14)15/h1-4,8H,5,11-12H2,(H,14,15)/t8-/m0/s1

InChI key

YGPSJZOEDVAXAB-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Key intermediate in the breakdown pathway of tryptophan.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue Jiang et al.
International journal of immunopathology and pharmacology, 34, 2058738420950584-2058738420950584 (2020-09-24)
Indoleamine 2,3-dioxygenase (IDO) was a potential tumor immunotherapy target. IDO inhibitors showed inconsistent results in clinical trials, but no preclinical comparative study was reported. The purpose of this study was to evaluate the differences of representative IDO inhibitors (PCC0208009, INCB024360
Ahmed Sadik et al.
Cell, 182(5), 1252-1270 (2020-08-21)
Aryl hydrocarbon receptor (AHR) activation by tryptophan (Trp) catabolites enhances tumor malignancy and suppresses anti-tumor immunity. The context specificity of AHR target genes has so far impeded systematic investigation of AHR activity and its upstream enzymes across human cancers. A

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service