Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

67540

Sigma-Aldrich

Methylhydrazine

purum, ≥98.0% (GC)

Synonym(s):

MMH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHNH2
CAS Number:
Molecular Weight:
46.07
Beilstein/REAXYS Number:
635645
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.6 (vs air)

Quality Level

vapor pressure

37.5 mmHg ( 20 °C)

grade

purum

assay

≥98.0% (GC)

autoignition temp.

385 °F

expl. lim.

97 %

refractive index

n20/D 1.4325 (lit.)
n20/D 1.434

bp

88-90 °C (lit.)

density

0.875 g/mL at 20 °C (lit.)

functional group

amine
hydrazine

SMILES string

CNN

InChI

1S/CH6N2/c1-3-2/h3H,2H2,1H3

InChI key

HDZGCSFEDULWCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methylhydrazine can be used:
  • In the synthesis of epoxides containing tetrasubstituted trifluoromethylated carbon center by the aerobic epoxidation of β,β-disubstituted enones.[1]
  • As a starting material along with 2-(methyl, phenyl or styryl)chromones for the synthesis of 3-(2-benzyloxy-6-hydroxyphenyl)-5-(methyl, phenyl or styryl)pyrazoles.[2]
  • As a starting material for one-step synthesis of [1,2]diazepino[4,5-b]indole derivatives by reacting with pyranoindolones.[3]

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

17.6 °F - closed cup

flash_point_c

-8 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-step synthesis of [1, 2] diazepino [4, 5-b] indole derivatives from the reaction of pyranoindolones with methylhydrazine
Hatzimimikou D, et al.
Synlett, 2008(12), 1773-1776 (2008)
R L Nelson et al.
American journal of surgery, 172(1), 85-88 (1996-07-01)
Selenium (SE) has been inversely associated with colon cancer risk. Two potential mechanisms of this effect were examined in a rodent short-term carcinogenesis assay: whether dietary SE deficiency altered the initiation aspect of carcinogenesis in the colon, and whether SE
M Gamberini et al.
Biochemical pharmacology, 45(9), 1913-1919 (1993-05-05)
The neutrophil-catalyzed metabolism of hydrazine derivatives to carbon-centered radicals was investigated by the spin-trapping technique using alpha-(4-pyridyl-1-oxide)-N-tert-butylnitrone (POBN). Oxidation of methylhydrazine (MeH), dimethylhydrazine (DMH), phenylethylhydrazine or procarbazine by neutrophils from rat peritoneal exudates led to the formation of alkyl radicals.
A Oubrie et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(21), 11787-11791 (1999-10-16)
Soluble glucose dehydrogenase (s-GDH) from the bacterium Acinetobacter calcoaceticus is a classical quinoprotein. It requires the cofactor pyrroloquinoline quinone (PQQ) to catalyze the oxidation of glucose to gluconolactone. The precise catalytic role of PQQ in s-GDH and several other PQQ-dependent
E S Lightcap et al.
Journal of medicinal chemistry, 39(3), 686-694 (1996-02-02)
(3-Hydroxybenzyl)hydrazine and methylhydrazine have been found to be potent slow-binding inhibitors of the pyridoxal 5-phosphate (PLP)-dependent enzyme gamma-aminobutyric acid aminotransferase (GABA-AT). Both compounds follow mechanism A (Morrison, J.F.; Walsh, C. T. Adv. Enzymol. 1988, 61, 201-301) which does not involve

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service