Skip to Content
MilliporeSigma

65280

1-Methoxy-2-propanol

ReagentPlus®, ≥99.0%

Synonym(s):

Propylene glycol methyl ether, Propyleneglycol monomethyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CH(OH)CH2OCH3
CAS Number:
Molecular Weight:
90.12
EC Number:
203-539-1
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1731270
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

3.12 (vs air)

vapor pressure

10.9 mmHg ( 25 °C)

product line

ReagentPlus®

assay

≥99.0%

autoignition temp.

532 °F

expl. lim.

13.8 %

refractive index

n20/D 1.403

bp

118-119 °C (lit.)

density

0.916 g/mL at 25 °C (lit.)

SMILES string

CC(O)COC

InChI

1S/C4H10O2/c1-4(5)3-6-2/h4-5H,3H2,1-2H3

InChI key

ARXJGSRGQADJSQ-UHFFFAOYSA-N

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under 1-Methoxy-2-propanol


pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

88.7 °F - closed cup - (External MSDS)

flash_point_c

31.5 °C - closed cup - (External MSDS)

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library