Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59210

Sigma-Aldrich

Isophthaloyl chloride

purum, ≥98.0% (AT)

Synonym(s):

Isophthaloyl dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(COCl)2
CAS Number:
Molecular Weight:
203.02
Beilstein/REAXYS Number:
638342
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.03 mmHg ( 25 °C)

Quality Level

grade

purum

assay

≥98.0% (AT)

form

solid

expl. lim.

8.9 %

bp

276 °C (lit.)

mp

43-44 °C (lit.)
43-47 °C

SMILES string

ClC(=O)c1cccc(c1)C(Cl)=O

InChI

1S/C8H4Cl2O2/c9-7(11)5-2-1-3-6(4-5)8(10)12/h1-4H

InChI key

FDQSRULYDNDXQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1A

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1

flash_point_f

356.0 °F - closed cup

flash_point_c

180 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pavle Mocilac et al.
Acta crystallographica. Section B, Structural science, 69(Pt 1), 62-69 (2013-02-01)
The one-step reaction of isophthaloyl dichloride with the 2-aminopyridine derivative (methyl 6-aminonicotinate) yields (i) a trimer-based macrocycle (EsIO)(3) and (ii) a tetramer-based macrocycle (EsIO)(4) in modest isolated synthetic yields (total of 25%), together with (iii) longer open-chain oligomers. The macrocyclization
[Experimental establishment of the MPEL for isophthaloyl chloride in reservoir water].
D G Deviatka et al.
Gigiena i sanitariia, (7)(7), 75-78 (1982-07-01)
Amir Abdolmaleki et al.
Amino acids, 42(5), 1997-2007 (2011-05-25)
N,N'-Bis[2-(methyl-3-(4-hydroxyphenyl)propanoate)]isophthaldiamide (5), a novel diol monomer containing chiral group, was prepared by the reaction of S-tyrosine methyl ester (3) with isophthaloyl dichloride (4a). A new family of optically active and potentially biodegradable poly(ester-amide)s (PEAs) based on tyrosine amino acid were
M Zhang et al.
Journal of microencapsulation, 12(4), 425-435 (1995-07-01)
Oil containing microcapsules were prepared by using phthaloyl dichloride as an oil-soluble monomer and diethylene triamine (DETA) as a water-soluble monomer. The diameter of the microcapsules decreased and their distribution became narrower and sharper as the emulsifying time was increased
Kira A Watkins et al.
International journal of pharmaceutics, 478(2), 496-503 (2014-12-10)
Hydrogels synthesized from poly(l-lysine isophthalamide) (PLP) crosslinked with l-lysine methyl ester were investigated as drug delivery systems for a wide size range of molecules (0.3-2000kDa). PLP is an anionic, pseudo-peptidic polymer that is an ideal hydrogel backbone due to its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service