Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56836

Sigma-Aldrich

MEGA-10 50 mM solution

Synonym(s):

MEGA-10 solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12161900

storage temp.

2-8°C

InChI

1S/C17H35NO6/c1-3-4-5-6-7-8-9-10-15(22)18(2)11-13(20)16(23)17(24)14(21)12-19/h13-14,16-17,19-21,23-24H,3-12H2,1-2H3/t13-,14+,16+,17+/m0/s1

InChI key

UMWKZHPREXJQGR-XOSAIJSUSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Mizuno et al.
Neurochemical research, 9(12), 1729-1735 (1984-12-01)
Receptors for benzodiazepines (BZ) and beta-carboline-carboxylic acid ethyl ester (beta-CCE) has been solubilized with decanoly-N-methylglucamide (DMG), a new kind of nonionic detergent. The apparent dissociation constants of diazepam and beta-CCE for solubilized receptor were similar to those for synaptic membranes.
H Kobayashi et al.
The Journal of veterinary medical science, 58(5), 477-479 (1996-05-01)
To prepare mycoplasmal DNA template for PCR from biological samples rapidly and easily, surfactants which can solubilize cell membranes effectively were investigated. 3-[3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate (CHAPS) was considered an effective surfactant. This surfactant could solubilize mycoplasma cell membrane without suppressing the polymerase
Shohei Nakamura et al.
Langmuir : the ACS journal of surfaces and colloids, 24(1), 15-18 (2007-12-07)
Solubilization of benzene, toluene, ethylbenzene, n-propylbenzene, n-butylbenzene, n-pentylbenzene, and n-hexylbenzene into micelles of decanoyl-N-methylglucamide (Mega-10) was studied, where equilibrium concentrations of the above solubilizates were determined spectrophotometrically at 303.2 K. The concentration of the above solubilizates remained constant below the
C Carnero Ruiz et al.
Biomacromolecules, 8(8), 2497-2503 (2007-07-17)
The protein-surfactant system constituted by bovine serum albumin (BSA) and N-decanoyl-N-methylglucamide (MEGA-10) has been studied by using surface tension, steady-state fluorescence, and dynamic light scattering measurements. It was found that the presence of protein delays the surfactant aggregation, which was
R Hovik et al.
The Biochemical journal, 290 ( Pt 1), 97-102 (1993-02-15)
The activity of the enzyme acyl-CoA oxidase (EC 1.3.99.3) is influenced by detergents. At concentrations above the critical micellar concentration, Triton X-100, Triton X-114 and Thesit stimulate oxidase activity. Lower concentrations of Triton X-100 and Triton X-114 render the acyl-CoA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service