54956
tert-Butyl (R)-2-hydroxybutyrate
purum, ≥98.0% (sum of enantiomers, GC)
About This Item
grade
purum
assay
≥98.0% (sum of enantiomers, GC)
form
solid
optical activity
[α]20/D +5.1±0.5°, c = 2% in carbon tetrachloride
optical purity
enantiomeric ratio: ≥99.5:0.5 (GC)
mp
50-54 °C
52-54 °C (lit.)
SMILES string
CC[C@@H](O)C(=O)OC(C)(C)C
InChI
1S/C8H16O3/c1-5-6(9)7(10)11-8(2,3)4/h6,9H,5H2,1-4H3/t6-/m1/s1
InChI key
UPRLOYNUXQZAPJ-ZCFIWIBFSA-N
Looking for similar products? Visit Product Comparison Guide
Application
replaced by
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service