Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

436984

Sigma-Aldrich

tert-Butyl (R)-2-hydroxybutyrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(OH)CO2C(CH3)3
CAS Number:
Molecular Weight:
160.21
Beilstein/REAXYS Number:
5728525
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

optical activity

[α]20/D +5.1°, c = 2 in carbon tetrachloride

optical purity

ee: 98% (GLC)

mp

52-54 °C (lit.)

SMILES string

CC[C@@H](O)C(=O)OC(C)(C)C

InChI

1S/C8H16O3/c1-5-6(9)7(10)11-8(2,3)4/h6,9H,5H2,1-4H3/t6-/m1/s1

InChI key

UPRLOYNUXQZAPJ-ZCFIWIBFSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service