Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52238

Supelco

Cetyl alcohol

for ion-selective electrodes, Selectophore, ≥99.0%

Synonym(s):

1-Hexadecanol, Palmityl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15OH
CAS Number:
Molecular Weight:
242.44
Beilstein/REAXYS Number:
1748475
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.05
grade:
for ion-selective electrodes
assay:
≥99.0% (GC)
≥99.0%
bp:
190 °C/15 mmHg (lit.)

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

assay

≥99.0% (GC)
≥99.0%

form

solid

bp

190 °C/15 mmHg (lit.)

mp

49-50 °C

SMILES string

CCCCCCCCCCCCCCCCO

InChI

1S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3

InChI key

BXWNKGSJHAJOGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cetyl alcohol may be used in hexadecyltrimethylammonium bromide-cetyl alcohol mixture for the emulsification of styrene.[1]
Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymer Colloids II (1980)
Luca Lo Piccolo et al.
Applied and environmental microbiology, 77(4), 1204-1213 (2010-12-25)
Enzymes involved in oxidation of long-chain n-alkanes are still not well known, especially those in gram-positive bacteria. This work describes the alkane degradation system of the n-alkane degrader actinobacterium Gordonia sp. strain SoCg, which is able to grow on n-alkanes
Flávia C Carvalho et al.
Journal of pharmaceutical sciences, 99(5), 2367-2374 (2009-12-08)
The development of a controlled-release dosage form of zidovudine (AZT) is of crucial importance, in view of the pharmacokinetics of its toxic activity. A suitable drug delivery system could increase AZT bioavailability, reducing its dose-dependent side effects. In this study
Dhanalakshmi S Anbukumar et al.
Journal of lipid research, 51(5), 1085-1092 (2009-12-19)
Neutrophils are important in the host response against invading pathogens. One chemical defense mechanism employed by neutrophils involves the production of myeloperoxidase (MPO)-derived HOCl. 2-Chlorohexadecanal (2-ClHDA) is a naturally occurring lipid product of HOCl targeting the vinyl ether bond of
Kerstin Wagner et al.
Chemistry and physics of lipids, 145(2), 119-127 (2006-12-26)
The monolayer structure of pure dipalmitoylphosphatidylcholine (DPPC) and equimolar mixtures of DPPC/n-hexadecanol (C(16)OH) and DPPC/dipalmitoylglycerol (DPG) are studied by the film balance technique and grazing incidence X-ray diffraction measurements. At 20 degrees C, the binary systems exhibit complete miscibility. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service