Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49760

Millipore

Gluten Hydrolysate from maize

suitable for microbiology

Synonym(s):

Peptone from corn

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41106212
NACRES:
NA.85

biological source

maize

Quality Level

form

powder

composition

total nitrogen (N), ≥11%

ign. residue

≤12%

loss

≤6.0% loss on drying

pH

5.0-6.0 (25 °C)

solubility

H2O: 2%, clear, brownish-yellow to brown

application(s)

food and beverages
microbiology

General description

An enzymatic digest of corn protein using non-animal enzymes of microbial or plant origin. Recommended for use in microbial media or as fermentation nutrient.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service