44479
2-Methylcyclohexanone
analytical standard
Synonym(s):
Tetrahydro-o-cresol
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥98.0% (GC)
shelf life
limited shelf life, expiry date on the label
refractive index
n20/D 1.447-1.449
n20/D 1.448 (lit.)
bp
162-163 °C (lit.)
density
0.924 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
SMILES string
CC1CCCCC1=O
InChI
1S/C7H12O/c1-6-4-2-3-5-7(6)8/h6H,2-5H2,1H3
InChI key
LFSAPCRASZRSKS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Flam. Liq. 3
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_c
48 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service