Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43340

Supelco

1,1,3,3-Tetramethyl-1,3-diphenyldisilazane

derivatization grade (GC derivatization), LiChropur

Synonym(s):

1,3-Diphenyl-1,1,3,3-tetramethyldisilazane, DPTMDS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H5Si(CH3)2]2NH
CAS Number:
Molecular Weight:
285.53
Beilstein/REAXYS Number:
2738118
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade (GC derivatization)

Quality Level

assay

≥97.0%

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.538 (lit.)
n20/D 1.538

bp

96-100 °C/0.1 mmHg (lit.)

density

0.985 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(N[Si](C)(C)c1ccccc1)c2ccccc2

InChI

1S/C16H23NSi2/c1-18(2,15-11-7-5-8-12-15)17-19(3,4)16-13-9-6-10-14-16/h5-14,17H,1-4H3

InChI key

HIMXYMYMHUAZLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,1,3,3-Tetramethyl-1,3-diphenyldisilazane (DPTMDS) is a derivatizing agent.

Application

DPTMDS may be used as ink to pattern semiconductor surfaces using Dip-Pen Nanolithography. It may also be used to deactivate the retention gap during GC-MS analysis of trimethylsilyl derivatives.

Other Notes

For the deactivation of glass capillary columns by persilylation ; The lithium amide is used for the selective formation of (Z)-enolates from ketones

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Grob et al.
Journal of Chromatography A, 244, 197-197 (1982)
Dip-pen? nanolithography on semiconductor surfaces.
Ivanisevic, Albena, and Chad A. Mirkin
Journal of the American Chemical Society, 123.32, 7887-7889 (2001)
P.A. McCarthy et al.
The Journal of Organic Chemistry, 52, 4681-4681 (1987)
Andréas Huenerbein et al.
Journal of chromatography. A, 985(1-2), 375-386 (2003-02-13)
The Medical Commission of the International Olympic Committee forbids the use of anabolic androgenic steroids and beta2-agonists to improve athletic performance. In this work we have selected examples of anabolic androgenic compounds and their metabolites to evaluate the GC-MS analysis
Concentration modulation by thermal decomposition for multiplex gas chromatography.
J R Valentin et al.
HRC & CC. Journal of high resolution chromatography & chromatography communications, 6, 621-622 (1983-11-01)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service