Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

428922

Supelco

Ammonium hydrogenphosphate, matrix modifier solution

~40 % (w/v) in H2O, 99.999%

Synonym(s):

Ammonium hydrogenphosphate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151816
PubChem Substance ID:
NACRES:
NA.21

assay

99.999%

concentration

~40 % (w/v) in H2O

density

1.203 g/mL at 25 °C

SMILES string

N.N.OP(O)(O)=O

InChI

1S/2H3N.H3O4P/c;;1-5(2,3)4/h2*1H3;(H3,1,2,3,4)

InChI key

MNNHAPBLZZVQHP-UHFFFAOYSA-N

General description

Matrix modification is a procedure done for reducing or even eliminating volatilization and vapour-phase interferences. This chemical modification is done by using inorganic salt solution which makes the interfering concomitant either more volatile or makes analyte less volatile. Ammonium hydrogen phosphate could be used in determination of lead to reduce the matrix effect.[1]

Application

For graphite furnace analyses

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Rodríguez García et al.
Talanta, 61(4), 509-517 (2008-10-31)
The occurrence of certain trace elements, such as cadmium, in honey bee can be considered as an indication of environmental pollution. In the present work, two methods for Cd determination in honey by electrothermal atomic absorption spectrometry, using palladium-magnesium nitrate
Lauri H. J. Lajunen, P. Peramaki
Spectrochemical Analysis by Atomic Absorption and Emission, 142-145 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service