Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41556

Supelco

Palatinose hydrate

analytical standard

Synonym(s):

6-O-α-D-Glucopyranosyl-D-fructose, Isomaltulose hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · xH2O
CAS Number:
Molecular Weight:
342.30 (anhydrous basis)
Beilstein/REAXYS Number:
93820
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

optical activity

[α]/D 97.0±3.0°, c = 1 in H2O

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

oligosaccharides

impurities

≤10% water

application(s)

food and beverages
personal care

format

neat

SMILES string

O.OC[C@H]1O[C@H](OC[C@H]2OC(O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11.H2O/c13-1-4-6(15)8(17)9(18)11(22-4)21-2-5-7(16)10(19)12(20,3-14)23-5;/h4-11,13-20H,1-3H2;1H2/t4-,5-,6-,7-,8+,9-,10+,11+,12?;/m1./s1

InChI key

XZKUCJJNNDINKX-HGLHLWFZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Palatinose is a disaccharide having an α(1→6) linkage between D-glucose and D-fructose and is similar to sucrose in its physicochemical properties. It is produced from sucrose in some bacteria by the action of sucrose isomerase. In mammalian gut, palatinose is hydrolyzed and absorbed much more slowly than sucrose.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service