Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40221

Sigma-Aldrich

Dimethyl disulfide

purum, ≥98.0% (GC)

Synonym(s):

DMDS, Methyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SSCH3
CAS Number:
Molecular Weight:
94.20
Beilstein/REAXYS Number:
1730824
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.24 (vs air)

vapor pressure

22 mmHg ( 20 °C)

grade

purum

assay

≥98.0% (GC)

autoignition temp.

>572 °F

expl. lim.

16 %

refractive index

n20/D 1.525 (lit.)
n20/D 1.527

bp

109 °C (lit.)

mp

−85 °C (lit.)

density

1.046 g/mL at 25 °C (lit.)

SMILES string

CSSC

InChI

1S/C2H6S2/c1-3-4-2/h1-2H3

InChI key

WQOXQRCZOLPYPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the α-methylsulfenylation of carbonyl compounds

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 1 Inhalation - STOT SE 3

target_organs

Central nervous system, Upper respiratory tract

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

59.0 °F - closed cup

flash_point_c

15 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhaodi Jiang et al.
Nature methods, 17(9), 937-946 (2020-08-12)
Genetically encoded tags for single-molecule imaging in electron microscopy (EM) are long-awaited. Here, we report an approach for directly synthesizing EM-visible gold nanoparticles (AuNPs) on cysteine-rich tags for single-molecule visualization in cells. We first uncovered an auto-nucleation suppression mechanism that
B.M. Trost
Chemical Reviews, 78, 363-363 (1978)
Chien-Jui Huang et al.
Pest management science, 68(9), 1306-1310 (2012-05-11)
Bacillus cereus C1L is a plant growth-promoting rhizobacterium and can elicit induced systemic resistance (ISR) in plants against necrotrophic pathogens. However, little is known about ISR elicitors produced by B. cereus C1L, and no ISR elicitor has been identified and
Anja Kristiansen et al.
Applied and environmental microbiology, 77(24), 8595-8604 (2011-10-18)
Biofiltration has proven an efficient tool for the elimination of volatile organic compounds (VOCs) and ammonia from livestock facilities, thereby reducing nuisance odors and ammonia emissions to the local environment. The active microbial communities comprising these filter biofilms have not
Gabrielle A Nevitt
Integrative and comparative biology, 51(5), 819-825 (2011-09-02)
Information transfer influences food-web dynamics in the marine environment, but infochemicals involved in these processes are only beginning to be understood. Dimethylsulfoniopropionate (DMSP) is produced by phytoplankton and other marine algae, and has been studied primarily in the context of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service