Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39584

Supelco

3-Sulfopyruvic acid lithium salt hydrate

≥97.0% (TLC)

Synonym(s):

2-Oxo-3-sulfopropanoic acid lithium salt hydrate, Lithium 3-sulfopyruvate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4O6S
Molecular Weight:
168.13
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (TLC)

impurities

≤15% water

format

neat

storage temp.

2-8°C

SMILES string

O=C(C(O)=O)CS(=O)(O)=O

InChI

1S/C3H4O6S/c4-2(3(5)6)1-10(7,8)9/h1H2,(H,5,6)(H,7,8,9)

InChI key

BUTHMSUEBYPMKJ-UHFFFAOYSA-N

Biochem/physiol Actions

3-Sulfopyruvic acid is the product of the transamination of cysteinesulfonate in a reaction catalyzed by aspartate amino- transferase. 3-Sulfopyruvic acid is stable and is reduced by malate dehydrogenase to beta-sulfolactate, which is excreted in the urine. Cysteinesulfonate, 3-sulfo- pyruvic acid, and beta-sulfolactate are reversibly interconverted in vivo.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C L Weinstein et al.
The Journal of biological chemistry, 263(8), 3735-3743 (1988-03-15)
L-Cysteinesulfonate (L-cysteate) is present in plasma, urine, and tissues in concentrations comparable to that of L-cysteinesulfinate, the primary oxidative metabolite of L-cysteine. Although cysteinesulfonate is known to be decarboxylated to taurine by cysteinesulfinate decarboxylase, the occurrence and importance of other
Mammalian sulfur amino acid metabolism: an overview.
O W Griffith
Methods in enzymology, 143, 366-376 (1987-01-01)
Three rare diseases in one Sib pair: RAI1, PCK1, GRIN2B mutations associated with Smith-Magenis Syndrome, cytosolic PEPCK deficiency and NMDA receptor glutamate insensitivity.
Adams DR, Yuan H, Holyoak T, et al.
Molecular Genetics and Metabolism, 113(3), 161-170 (2014)
Signaling And Metabolism In Cancer: Endocrine Pancreas Deficiency And Hybrid Anabolism - Catabolism, Drugs That Undo The Process
Israel, M.
Cancer Therapy, 10, 1-1 (2014)
Ines Thiele et al.
Nature biotechnology, 31(5), 419-425 (2013-03-05)
Multiple models of human metabolism have been reconstructed, but each represents only a subset of our knowledge. Here we describe Recon 2, a community-driven, consensus 'metabolic reconstruction', which is the most comprehensive representation of human metabolism that is applicable to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service