Skip to Content
MilliporeSigma

34269

2,6-Dibromophenol

purum, ≥97.0% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Br2C6H3OH
CAS Number:
Molecular Weight:
251.90
EC Number:
210-161-0
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
2043614
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥97.0% (HPLC)

bp

255-256 °C/740 mmHg (lit.)

mp

53-56 °C

SMILES string

Oc1c(Br)cccc1Br

InChI

1S/C6H4Br2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9H

InChI key

SSIZLKDLDKIHEV-UHFFFAOYSA-N

Gene Information



Still not finding the right product?

Explore all of our products under 2,6-Dibromophenol




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library