Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32943

Supelco

Deoxynivalenol

Deoxynivalenol

reference material

Synonym(s):

3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, Vomitoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O6
CAS Number:
Molecular Weight:
296.32
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
reference material

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

[H][C@]12O[C@]3([H])[C@H](O)C[C@@](C)([C@]34CO4)[C@@]1(CO)[C@H](O)C(=O)C(C)=C2

InChI

1S/C15H20O6/c1-7-3-9-14(5-16,11(19)10(7)18)13(2)4-8(17)12(21-9)15(13)6-20-15/h3,8-9,11-12,16-17,19H,4-6H2,1-2H3/t8-,9-,11-,12-,13-,14-,15+/m1/s1

InChI key

LINOMUASTDIRTM-QGRHZQQGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Deoxynivalenol belongs to the class of type B trichothecene mycotoxins typically produced by the species of Fusarium genus.[1] It is cytotoxic and is known to hinder the macromolecular synthesis.[2]

Application

Deoxynivalenol may be used as a reference standard for the determination of the analyte in:
  • Cereals by high performance liquid chromatography-mass spectrometry (HPLC-MS),[1] ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)[3] and HPLC-MS/MS.[4]
  • Biological fluidsby HPLC coupled to diode array detector (DAD)[5]and LC-MS/MS.[6]
  • Eggs by HPLC with ultraviolet (UV) detection.[7]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of deoxynivalenol and de-epoxy-deoxynivalenol in animal tissues by liquid chromatography after clean-up with an immunoaffinity column.
Valenta H, et al.
Mycotoxin Research, 19(1), 51-51 (2003)
Masked mycotoxins: determination of a deoxynivalenol glucoside in artificially and naturally contaminated wheat by liquid chromatography- tandem mass spectrometry.
Berthiller F, et al.
Journal of Agricultural and Food Chemistry, 53(9), 3421-3425 (2005)
Quantitative determination of T-2 toxin, HT-2 toxin, deoxynivalenol and deepoxy-deoxynivalenol in animal body fluids using LC-MS/MS detection.
De Baere S, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 879(24), 2403-2415 (2011)
Study on the transmission of deoxynivalenol and de-epoxy-deoxynivalenol into eggs of laying hens using a high-performance liquid chromatography-ultraviolet method with clean-up by immunoaffinity columns.
Valenta H and Danicke S
Molecular Nutrition And Food Research, 49(8), 779-785 (2005)
Determination of nivalenol and deoxynivalenol in wheat using liquid chromatography-mass spectrometry with negative ion atmospheric pressure chemical ionisation.
Razzazi-Fazeli E, et al.
Journal of Chromatography A, 854(1-2), 45-55 (1999)

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service