
Deoxynivalenol

Synonym(s):
3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, Vomitoxin
Empirical Formula (Hill Notation):
C15H20O6
CAS Number:
Molecular Weight:
296.32
Products
Product Category
Brand
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing