32863
4-(3,6-Dimethyl-3-heptyl)phenol monoethoxylate solution
10 μg/mL in acetone, analytical standard
Synonym(s):
2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy]ethanol, 2-[4-(3,6-Dimethyl-3-heptyl)phenoxy]ethanol, 3,6,3-Nonylphenol-monoethoxylate, 363-NP1EO
About This Item
Recommended Products
grade
analytical standard
shelf life
limited shelf life, expiry date on the label
concentration
10 μg/mL in acetone
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
single component solution
storage temp.
−20°C
SMILES string
CCC(C)(CCC(C)C)c1ccc(OCCO)cc1
InChI
1S/C17H28O2/c1-5-17(4,11-10-14(2)3)15-6-8-16(9-7-15)19-13-12-18/h6-9,14,18H,5,10-13H2,1-4H3
InChI key
QRFCMJANHDRHEH-UHFFFAOYSA-N
General description
Application
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
1.4 °F
flash_point_c
-17 °C
ppe
Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service