32591
Baquiloprim
VETRANAL®, analytical standard
Synonym(s):
5-{[8-(Dimethylamino)-7-methyl-5-quinolinyl]methyl}-2,4-pyrimidinediamine
Select a Size
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
forensics and toxicology
pharmaceutical (small molecule)
format
neat
SMILES string
CN(C)c1c(C)cc(Cc2cnc(N)nc2N)c3cccnc13
InChI
1S/C17H20N6/c1-10-7-11(8-12-9-21-17(19)22-16(12)18)13-5-4-6-20-14(13)15(10)23(2)3/h4-7,9H,8H2,1-3H3,(H4,18,19,21,22)
InChI key
AIOWJIMWVFWROP-UHFFFAOYSA-N
General description
Application
- Bovine muscle samples by solid-phase extraction (SPE) and hydrophilic interaction liquid chromatography–tandem mass spectrometry (HILIC-MS/MS).[2]
- Sewage sludge samples by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction and liquid chromatography coupled to time-of-flight mass spectrometry (LC-TOF-MS).[3]
Recommended products
Legal Information
related product
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service