Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

291870

Sigma-Aldrich

3-Noradamantanamine hydrochloride

95%

Synonym(s):

3-Aminonoradamantane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N · HCl
CAS Number:
Molecular Weight:
173.68
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

assay

95%

form

solid

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl.N[C@@]12C[C@H]3C[C@H](C[C@H]1C3)C2

InChI

1S/C9H15N.ClH/c10-9-4-6-1-7(5-9)3-8(9)2-6;/h6-8H,1-5,10H2;1H/t6-,7+,8-,9-;

InChI key

YPEGOYFGBDTDJK-OAGGSKBVSA-N

Application

Building block for "Anti-Bredt"strained molecules.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 18, 82-82 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service