Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28001

Supelco

Boscalid Metabolite M510F01

analytical standard

Synonym(s):

2-Chloro-N-(4′-chloro-5-hydroxybiphenyl-2-yl)nicotinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12Cl2N2O2
CAS Number:
Molecular Weight:
359.21
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

~5% water (calc. from elemental analysis)

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=NC=CC=C1C(NC2=C(C3=CC=C(Cl)C=C3)C=C(O)C=C2)=O

General description

M510F01 is the hydroxylated metabolite of the agrochemical, boscalid,[1] mainly developed for the protection of crops in agriculture. It is a succinate dehydrogenase inhibitor (SDHI) belonging to the anilide group of fungicides.[2]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pesticide residues in food 2019 - Report 2019: Extra Joint FAO/WHO Meeting on Pesticide Residues 2019 (2019)
Francesc A Esteve-Turrillas et al.
Food chemistry, 267, 2-9 (2018-06-24)
Boscalid is an agrochemical recently developed for crop protection and the most significant member of the succinate dehydrogenase inhibitor group of fungicides. In this study, a collection of high-affinity monoclonal antibodies was generated to boscalid. By using a series of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service