Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

258113

Sigma-Aldrich

Nitric acid

ACS reagent, 70%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

1 (vs air)

vapor pressure

8 mmHg ( 20 °C)

assay

68.0-70.0% (ACS specification)
70%

ign. residue

≤5 ppm

color

APHA: ≤10

bp

120.5 °C (lit.)

density

1.413 g/mL at 25 °C

anion traces

chloride (Cl-): ≤0.5 ppm
sulfate (SO42-): ≤1 ppm

cation traces

As: ≤0.01 ppm
Fe: ≤0.2 ppm
heavy metals (as Pb): ≤0.2 ppm

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibromomethane-d2 is a deuterated NMR solvent that is useful in NMR-based research and analyses.

Application

Dibromomethane-d2 may be used in the synthesis [18F]?uoromethylbromide-d2. This compound is used for the alkylation of N-(3,5-dichlorobenzyl)-4-hydroxybenzenecarboximidamide to form a ?uorine-18-labelled tracer that can be used in PET imaging studies.[1]

Other Notes

This product has been replaced by 695033-Sigma-Aldrich | Nitric acid
ACS reagent, 70%

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

supp_hazards

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis of a benzamidine?containing NR2B?selective NMDA receptor ligand labelled with tritium or fluorine?18.
Hamill TG, et al
Journal of Labelled Compounds & Radiopharmaceuticals, 48(1), 1-10 (2005)
Nicole McGowan et al.
The Science of the total environment, 494-495, 177-186 (2014-07-22)
Microchemical analysis of otolith (calcified 'ear stones' used for balance and orientation) of fishes is an important tool for studying their environmental history and management. However, the spatial resolution achieved is often too coarse to examine short-term events occurring in
Alyson M Baergen et al.
Environmental science & technology, 47(2), 815-820 (2012-12-15)
The fate of NO(x) (=NO + NO(2)) is important to understand because NO(x) is a significant player in air quality determination through its role in O(3) formation. Here we show that renoxification of the urban atmosphere may occur through the
Lauren E Kremer et al.
Journal of inorganic biochemistry, 147, 227-234 (2015-05-11)
Uncontrolled reactions in biological media are a main obstacle for clinical translation of V-based anti-diabetic or anti-cancer pro-drugs. We investigated the use of controlled-release pharmaceutical formulations to ameliorate this issue with a series of V(V) and (IV) complexes of anionic
Marie-Pierre Isaure et al.
Journal of experimental botany, 66(11), 3201-3214 (2015-04-16)
Arabidopsis halleri is a model plant for Zn and Cd hyperaccumulation. The objective of this study was to determine the relationship between the chemical forms of Cd, its distribution in leaves, and Cd accumulation and tolerance. An interspecific cross was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service