Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21144

Sigma-Aldrich

Calcium D-gluconate monohydrate

tested according to Ph. Eur.

Synonym(s):

Calcii gluconas, D-Gluconic acid calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22CaO14 · H2O
CAS Number:
Molecular Weight:
448.39
Beilstein/REAXYS Number:
3919646
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

agency

USP/NF
tested according to Ph. Eur.

Quality Level

form

powder

color

white

cation traces

(MAGNESIUM AND ALKALI METALS MAX. 0.4% (2 MG))

storage temp.

room temp

SMILES string

O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O[Ca]OC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/2C6H12O7.Ca.H2O/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;;/h2*2-5,7-11H,1H2,(H,12,13);;1H2/q;;+2;/p-2/t2*2-,3-,4+,5-;;/m11../s1

InChI key

XLNFVCRGJZBQGX-XRDLMGPZSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Calcium D-gluconate monohydrate has been used as a soluble precursor for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture.[1]

Biochem/physiol Actions

Calcium gluconate exhibits protective impact against osteoarthritis (OA) in Sprague-Dawley rats by inhibiting COX-2 as well as other related chondrocyte apoptosis.[2] It also facilitates the excision of skin necrosis by versajet hydrosurgery due to calcium gluconate extravasation in pediatric patients.[3]

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su-Jin Kang et al.
Journal of orthopaedic surgery and research, 9(1), 14-14 (2014-03-08)
This study aimed to determine whether calcium gluconate exerts protective effects on osteoarthritis (OA) induced by anterior cruciate ligament (ACL) transection and partial medial meniscectomy. Calcium gluconate was administered by mouth daily for 84 days to male ACL transected and
Nazan Sivrioğlu et al.
Acta orthopaedica et traumatologica turcica, 48(1), 6-9 (2014-03-20)
The aim of the study was to evaluate the results of debridement with Versajet hydrosurgery system in patients with skin necrosis after ca gluconate extravasation. We evaluated nine infants (mean age: 26 days; range: 1 day to 3 months) with
Changlei Qin et al.
Environmental science & technology, 46(3), 1932-1939 (2012-01-06)
Three types of sorbent were fabricated using various calcium and support precursors via a simple mixing method, in order to develop highly effective, durable, and cheap CaO-based sorbents suitable for CO(2) capture. The sorption performance and morphology of the sorbents

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service