Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

207195

Supelco

Indium atomic absorption standard solution

analytical standard, 1,000 μg/mL In in 1 wt. % HNO3 (Exact In concentration on label)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

concentration

1,000 μg/mL In in 1 wt. % HNO3 (Exact In concentration on label)

technique(s)

AAS: suitable

density

1.01 g/mL at 25 °C

format

single component solution

pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service