Skip to Content
MilliporeSigma

16320

2-Bromobenzoic acid

purum, ≥95.0% (HPLC)

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
BrC6H4CO2H
CAS Number:
Molecular Weight:
201.02
EC Number:
201-848-6
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
971266
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI key

XRXMNWGCKISMOH-UHFFFAOYSA-N

InChI

1S/C7H5BrO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4H,(H,9,10)

SMILES string

OC(=O)c1ccccc1Br

vapor pressure

>1 mmHg ( 20 °C)

grade

purum

assay

≥95.0% (HPLC)

mp

147-150 °C (lit.)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K H Engesser et al.
FEMS microbiology letters, 51(1), 143-147 (1989-07-15)
Pseudomonas putida strain CLB 250 (DSM 5232) utilized 2-bromo-, 2-chloro- and 2-fluorobenzoate as sole source of carbon and energy. Degradation is suggested to be initiated by a dioxygenase liberating halide in the first catabolic step. After decarboxylation and rearomatization catechol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service